Order Entry
Austria
ContactUsLinkComponent
Glass beads, MP Biomedicals
Glass beads, MP Biomedicals
Catalog # ICNA116914801
Supplier:  MP Biomedicals
Glass beads, MP Biomedicals
Catalog # ICNA116914801
Supplier:  MP Biomedicals

Specifications

  • Ø:
    4 mm
  • Description:
    Glass beads
  • Pk:
    100

Specifications

About this item

Glass beads.

  • For research use only
  • Can be stored at 2 to 8 °C

Glass beads used as packing for distillation columns, mixing beads and boiling stones.