Order Entry
Austria
ContactUsLinkComponent
Genomic DNA purification kits, Fast DNA™ SPIN kit for Soil, MP Biomedicals
Genomic DNA purification kits, Fast DNA™ SPIN kit for Soil, MP Biomedicals
Catalog # ICNA116560600
Supplier:  MP Biomedicals
CAS Number:  
Genomic DNA purification kits, Fast DNA™ SPIN kit for Soil, MP Biomedicals
Catalog # ICNA116560600
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    10 Tests
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Stool
    Wastewater
    Soil
  • Target molecule:
    DNA
  • Description:
    Fast DNA™ SPIN Kit for Soil, 50 ml
  • Description:
    Fast DNA™ SPIN Kit for Soil

Specifications

About this item

The FastDNA™ SPIN Kit for Soil is designed to efficiently isolate bacterial, fungi, plant and animal genomic DNA from soil and environmental samples.

Up to 5 g of soil are processed by the FastPrep®-24 or FastPrep®-96 instrument with the Lysing Matrix E tubes designed to efficiently lyse all microorganisms, including difficult sources such as eubacterial spores and endospores, gram positive bacteria and yeast. The released DNA is purified by a silica-based spin filter columns and removes humic acids/polyphenols that can inhibit PCR. The resulting pure DNA is suitable for PCR analysis and other downstream applications.

Delivery: Garnet lysing matrix tubes (50 ml), 10 each; PLS solution, 60 ml; MT buffer, 15 ml; sodium phosphate buffer, 350 ml; PPS for soil, 25 ml; binding matrix, 120 ml; GTC wash buffer, 120 ml; SEWS-M, 12 ml; TES, 60 ml; BBS gel loading dye, 200 µl; SPIN filter tubes (50 ml), 10 each.