Brilliant cresyl blue solution biological stain, for microscopy, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:81029-05-2
:10127-36-3
Formula:C₃₄H₄₂Cl₄N₈O₂Zn
MDL Number:MFCD00148901
:801.96 g/mol
Density:1.01 g/cm3 (20 °C)
Storage Temperature:Ambient
:1
Specification Test Results
- Application test (Reticulocyte staining):passes test
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.01384.0100_TDS_EN
Product specification
Merck The Color of Tissue Diagnostics
Product Literature
msds_101384
SDS
Frequently Bought Together