ContactUsLinkComponent
VWR® pHenomenal® Redox Electrode
VWR® pHenomenal® Redox Electrode
  662-1165
undefined
VWR® pHenomenal® Redox Electrode
  662-1165
  • Connection:
    BNC, 1 m cable
  • Description:
    Redox electrode pHenomenal® ORP220, refillable
  • Material:
    Glass
  • Electrolyte:
    3 M KCl
  • Pk:
    1
Redox electrode with BNC connector and cable.
 : Includes pH-electrode only.