ContactUsLinkComponent
Reusable earplugs, 1291, E-A-R™ Ultrafit™
Reusable earplugs, 1291, E-A-R™ Ultrafit™
Catalog # 111-0450
Supplier:  3M
undefined
Reusable earplugs, 1291, E-A-R™ Ultrafit™
Catalog # 111-0450
Supplier:  3M
Specifications
  • Colour:
    Yellow
  • Description:
    Earplugs, with cord
  • Size:
    U*
  • Pk:
    50 PAIR
Specifications
About this item
These earplugs are made from flexible materials that are already pre-formed to ensure optimum fit.
  • The triple-flange feature of the earplugs ensures a perfect fit: No size adjustment required
  • Easy to insert
  • Washable and reusable
SNR: 32 dB
Certifications: EN 352-2
Caution: * One size fits all.