Order Entry
Austria
ContactUsLinkComponent
Chlorine analyser, ChloroSense®
Chlorine analyser, ChloroSense®
Catalog # 705-0650
Supplier:  Palintest
undefined
Chlorine analyser, ChloroSense®
Catalog # 705-0650
Supplier:  Palintest

Specifications

  • Model:
    ChloroSense®
  • Range:
    Up to 10 mg/l
  • Resolution:
    Free chlorine: 0,01 mg/l @ 1 mg/l ; Total chlorine: 0,1 mg/l @ 10 mg/l
  • Temperature range:
    2...40 °C (instrument)
    2...25 °C (probe)
  • Description:
    Analyser for chlorine testing
  • Temperature resolution:
    0,1 °C
  • Batteries:
    4× AA
  • Display:
    LCD (42×22 mm)
  • Measurement time:
    55 s
  • Pk:
    1 KIT

Specifications

About this item

The ChloroSense is designed to determine free and total chlorine in less than one minute.

  • Alternative to the DPD method ((N,N-diethylparaphenylenediamine)
  • Reagent-free and operator independent method
  • Patented sensor for fast and accurate results

Designed for drinking water, it is used in other applications including washing of food produce, testing of potable water storage tanks, pipe refurbishment as well as in hospitals.

Delivery: Supplied with electrodes, calibration chip, sample bottles and carrying case.