Avantor® Apollo C18, HPLC Columns
Supplier: Avantor
Certificates
About this item
Avantor® Apollo HPLC columns use high purity, base deactivated silica for powerful separations.
- Easy scale-up from analytical to preparative
- Extended pH stability is about 1,5 to 10,5
- USP L1
These columns are ideal for routine analysis and for educational laboratories.
Specifications
- Catalog No:
- HICH970205
- HICH36521
- HICH36537
- HICH36543
- HICH36515
- 96454/N
- HICH36511
- HICH36505
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Specifications
Guard holders and accessories
Product Information
Frequently Bought Together