Order Entry
Austria
ContactUsLinkComponent
Sodium pyruvate solution 100 mM with 8.5 g/L NaCl cell culture reagent, Corning®
Sodium pyruvate solution 100 mM with 8.5 g/L NaCl cell culture reagent, Corning®
Catalog # 25-000-CI
Supplier:  Corning
CAS Number:  
undefined
Sodium pyruvate solution 100 mM with 8.5 g/L NaCl cell culture reagent, Corning®
Catalog # 25-000-CI
Supplier:  Corning
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    6
  • Seq:
    53
  • Pk Info:
    6 x 100 ml

Specifications

Specification Test Results

  • pH:
    6.0 ± 1.0
  • Osmolality:
    400 ± 100 mOsm/Kg H20
  • Endotoxin:
    < 0.25 EU/ml@ 1:100
  • Mycoplasma:
    Tested Negative
  • Sterility :
    Pass
  • Cell Growth:
    Pass

Specification Test Results

About this item

 :
 :110.04 g/mol
Storage Temperature:Refrigerator
CAS Number:113-24-6
Formula:CH₃COCOONa
MDL Number:MFCD00002586