Order Entry
Austria
ContactUsLinkComponent
L(+)-Glutamine solution 200 mM in water (29.20 mg/ml) cell culture reagent, Corning®
L(+)-Glutamine solution 200 mM in water (29.20 mg/ml) cell culture reagent, Corning®
Catalog # 25-005-CI
Supplier:  Corning
CAS Number:  
undefined
L(+)-Glutamine solution 200 mM in water (29.20 mg/ml) cell culture reagent, Corning®
Catalog # 25-005-CI
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    6
  • Seq:
    0006
  • Pk Info:
    6×100 ml

Specifications

Specification Test Results

  • pH:
    5.5 ± 0.5
  • Osmolality:
    450 ± 50 mOsm/kg H2O
  • Endotoxin:
    < 0.25 EU/ml @ 1 :50
  • Mycoplasma:
    Tested Negative
  • Sterility :
    Pass
  • Cell Growth:
    Pass

Specification Test Results

About this item

 :
 :146.15 g/mol
CAS Number:56-85-9
Formula:C₅H₁₀N₂O₃
MDL Number:MFCD00008044