Spin module and recovery tubes for DNA purification, MP Biomedicals
Spin module and recovery tubes are used to separate particles from solutions in a few seconds. They are designed for optimal performance, providing faster and cleaner separations than traditional methods, such as columns, filters, or simple centrifugation.
- Fast separation
- Pure DNA
- No matrix carry over
When used with the GENECLEAN® or MERmaid® protocols, they combine the speed, efficiency, and selectivity of the GLASSMILK® binding reaction with an even more rapid execution of the separation, washing and elution steps.
- Sample type:DNA preps
- Target molecule:DNA
- Sample size:500-700 µL liquid per centrifugation; Larger volumes require multiple loadings
Frequently Bought Together