Plasmid purification, MiniPrep Express™ Matrix, MP Biomedicals
Catalog # ICNA112000200
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:1 KIT
- Method / Format:Precipitation
- Prep size:Mini
- Refine target molecule:Plasmid
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:MiniPrep Express™ Matrix, sufficient for 1250 preps
- Description:MiniPrep Express™ Matrix
Specifications
About this item
MiniPrep Express™ Matrix is designed for rapid purification of plasmid DNA. It is an ideal tool for screening hundreds of recombinant clones in record time using restriction enzyme analysis and PCR.
MiniPrep Express™ Matrix provides the essential ingredients for 'make your own' miniprep kits for rapid screening that involve a restriction digest. This kit allows you to make the lysis reagents required for the most commonly used methods.
Delivery information: Kit contains MiniPrep Express™ Matrix, protocols and recipes necessary for either alkaline lysis or boiling miniprep procedures.