Order Entry
Austria
ContactUsLinkComponent
 
RNA Purification kits, RNAID® spin kit, MP Biomedicals
  ICNA111107200
 :  
RNA Purification kits, RNAID® spin kit, MP Biomedicals
  ICNA111107200
 :  

 

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    RNA gel bands
  • Sample type:
    Gel matrix
  • Target molecule:
    RNA
  • Description:
    RNaid® Spin Kit
  • Description:
    RNaid® Spin Kit

 

 

The RNaid® Spin Kit is designed to isolate and purify RNA (75 bases to 10 kb) from native and denaturing agarose and polyacrylamide gels. The kit includes spin filters which prevent silica particle carry-over into cleaned RNA.

Ideal for removal of unincorporated nucleotides, buffer salts, and other reagents from enzyme reactions. The purified RNA is ready for reverse transcription and PCR, labeling and microarray analysis, and other common applications.