Order Entry
Austria
ContactUsLinkComponent
DNA purification kits, Geneclean® kit, MP Biomedicals
DNA purification kits, Geneclean® kit, MP Biomedicals
Catalog # ICNA111001200
Supplier:  MP Biomedicals
CAS Number:  
undefined
DNA purification kits, Geneclean® kit, MP Biomedicals
Catalog # ICNA111001200
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    DNA gel bands
    Amplified/modified DNA
  • Sample type:
    Gel matrix
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    GeneClean® Kit
  • Description:
    GeneClean® Kit

Specifications

About this item

The GeneClean® Kit is designed to purify fragments of DNA from 200 bp to 20 kb from solutions and agarose gels.

Applications include desalting, elimination of proteins from enzymatic reactions, removal of primers and unincorporated nucleotides from enzymatic reactions, separation of linearised from uncut vector, and isolation of PCR products away from genomic DNA.

The kit simplifies the process of purifying DNA into three easy steps: Bind, wash and elute. Ethanol precipitation is never required and purified DNA is immediately ready for a wide variety of downstream applications.

Accessories information: Geneclean® Glassmilk™ is a specially prepared aqueous suspension of proprietary silica matrix that allows flexibility of reaction size and volume. Geneclean® Glassmilk™ binds to radioactive isotope-labeled ssDNA or dsDNA purified from agarose resulting in purified DNA suitable for further use.
Delivery: Supplied with Geneclean® Glassmilk™, which is also available separately.