Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Cell Culture Buffers
Hank's balanced salt solution, modified, MP Biomedicals
Hank's balanced salt solution, modified, MP Biomedicals
Hank's balanced salt solution, modified, MP Biomedicals
Catalog # ICNA091810054
Supplier:
MP Biomedicals
CAS Number:
Expand image
Hank's balanced salt solution, modified, MP Biomedicals
Catalog # ICNA091810054
Supplier:
MP Biomedicals
CAS Number:
Specifications
Pk:
500 mL
Description:
Hank's balanced salt solution, modified (1X)
Specifications