Order Entry
Austria
ContactUsLinkComponent
Cleanroom mops, SilberKing
Cleanroom mops, SilberKing
  PPSA2160000
undefined
Cleanroom mops, SilberKing
  PPSA2160000

 

  • Description:
    SilberKing, 150 g
  • Length:
    400 mm
  • Pk:
    1

 

 

Mops for all hygienically sensitive areas, buildings without dryer.

  • Suitable in areas in which mops need to be stored for longer times
  • For impregnation of mop covers without disinfectants
  • Ready-to-use pre-washed
  • Made up of synthetic/viscose yarn (50:50)
  • With cover sheet, silver, PVC pocket and sewed up double-latching pocket