Wipe rolls, WYPALL* L10 EXTRA+
Lieferant: KIMBERLY CLARK
Zertifikate
Über diesen Artikel
These single use wipes made from single ply AIRFLEX* material are ideal for multipurpose cleaning and medium duty wiping tasks such as polishing glass, smooth surfaces and cleaning tools.
- Stronger than conventional single ply wipes
- Strong and absorbent
- Clean up fast without disintegrating
- Can be used with a dispensing system to help control use and reduce wastage
Large rolls: For use with dispensers 6146, 6154 and 6155.
Centrefeed rolls: For use with dispensers 7017 and 7087.
Small rolls: For use with dispenser 7041.
Roll Control wipes: For use with dispensers 7018 and 7181.
Frequently Bought Together