Orders LinkContactUsLinkComponent
Sample loops for sample injectors, Rheodyne®
Supplier:   IDEX RHEODYNE
Sample loops for sample injectors, Rheodyne®
Supplier:  IDEX RHEODYNE
Certificates
Sample loops for sample injectors, Rheodyne®
Sample loops for sample injectors, Rheodyne®
35 Options Available Below
Product Details & Documents
Product Family Options
Sample loops, for preparative Rheodyne® injectors
Product Information
  • Material
    Capacity
    For tube I-Ø
    For tube O-Ø
    Availability
    Price
  • 554-1536
    Sample loop
    Sample loop
    PEEK
    10 ml
    2,0 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 590,00 €
    590,00 €
    1 * 1 ST
       590,00 €
  • 554-1537
    Sample loop
    Sample loop
    PEEK
    20 ml
    2,0 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 800,00 €
    800,00 €
    1 * 1 ST
       800,00 €
  • 554-1539
    Sample loop
    Sample loop
    Stainless steel
    5 ml
    2,0 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 589,00 €
    589,00 €
    1 * 1 ST
       589,00 €
  • 554-1540
    Sample loop
    Sample loop
    Stainless steel
    10 ml
    2,0 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 814,00 €
    814,00 €
    1 * 1 ST
       814,00 €
  • 554-1541
    Sample loop
    Sample loop
    Stainless steel
    20 ml
    2,0 mm
    3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 210,00 €
    1 210,00 €
    1 * 1 ST
       1 210,00 €
Sample loops, for analytical Rheodyne® injectors
Product Information
  • Material
    Capacity
    For tube I-Ø
    For tube O-Ø
    Availability
    Price
  • 554-2154
    Sample loop, for model 7725, 7725i, 9725(i)
    Sample loop, for model 7725, 7725i, 9725(i)
    PEEK
    2 µl
    — mm
    — mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 338,00 €
    338,00 €
    1 * 1 ST
       338,00 €
  • 554-2155
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Stainless steel
    5 µl
    0,18 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 125,00 €
    125,00 €
    1 * 1 ST
       125,00 €
  • 554-2176
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    PEEK
    5 µl
    0,18 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 93,20 €
    93,20 €
    1 * 1 ST
       93,20 €
  • 554-2156
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Stainless steel
    10 µl
    0,30 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 119,00 €
    119,00 €
    1 * 1 ST
       119,00 €
  • 554-2177
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    PEEK
    10 µl
    0,30 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 116,00 €
    116,00 €
    1 * 1 ST
       116,00 €
  • 554-2157
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Stainless steel
    20 µl
    0,30 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 110,00 €
    110,00 €
    1 * 1 ST
       110,00 €
  • 554-2178
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    PEEK
    20 µl
    0,51 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 114,00 €
    114,00 €
    1 * 1 ST
       114,00 €
  • 554-2158
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Stainless steel
    50 µl
    0,51 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 120,00 €
    120,00 €
    1 * 1 ST
       120,00 €
  • 554-2179
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    PEEK
    50 µl
    0,51 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 114,00 €
    114,00 €
    1 * 1 ST
       114,00 €
  • 554-2180
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    PEEK
    100 µl
    0,51 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 114,00 €
    114,00 €
    1 * 1 ST
       114,00 €
  • 554-2160
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Stainless steel
    200 µl
    0,76 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €
  • 554-2181
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    PEEK
    200 µl
    0,76 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 157,00 €
    157,00 €
    1 * 1 ST
       157,00 €
  • 554-1548
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Stainless steel
    500 µl
    0,76 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 145,00 €
    145,00 €
    1 * 1 ST
       145,00 €
  • 554-2182
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    PEEK
    500 µl
    0,76 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 189,00 €
    189,00 €
    1 * 1 ST
       189,00 €
  • 554-2159
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Stainless steel
    1 ml
    0,51 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 125,00 €
    125,00 €
    1 * 1 ST
       125,00 €
  • 554-2161
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Stainless steel
    1 ml
    0,76 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 168,00 €
    168,00 €
    1 * 1 ST
       168,00 €
  • 554-2183
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    PEEK
    1 ml
    0,76 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 211,00 €
    211,00 €
    1 * 1 ST
       211,00 €
  • 554-2162
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Stainless steel
    2 ml
    1,00 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 245,00 €
    245,00 €
    1 * 1 ST
       245,00 €
  • 554-2184
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    PEEK
    2 ml
    1,00 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 302,00 €
    302,00 €
    1 * 1 ST
       302,00 €
  • 554-2185
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    Sample loop, for model 9725, 9010, PR/EV750-100, PR/EV753-100
    PEEK
    5 ml
    1,00 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 550,00 €
    550,00 €
    1 * 1 ST
       550,00 €
  • 554-2163
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Sample loop, for model 7725, 7725I, PR/EV 700-100, PR/EV 703-100
    Stainless steel
    5 ml
    1,00 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 420,00 €
    420,00 €
    1 * 1 ST
       420,00 €
Sample loops, for Micro Rheodyne® injectors
Product Information
  • Material
    Capacity
    For tube I-Ø
    For tube O-Ø
    Availability
    Price
  • 554-2165
    Sample loop, for model 8125
    Sample loop, for model 8125
    Stainless steel
    0,01 ml
    0,20 mm
    0,508 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 206,00 €
    206,00 €
    1 * 1 ST
       206,00 €
Sample loops, for syringe loading Rheodyne® injectors
Product Information
  • Material
    Capacity
    For tube I-Ø
    For tube O-Ø
    Availability
    Price
  • 554-2069
    Sample loop, for model 7125, 7010
    Sample loop, for model 7125, 7010
    Stainless steel
    5 µl
    0,18 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 127,00 €
    127,00 €
    1 * 1 ST
       127,00 €
  • 554-2070
    Sample loop, for model 7125, 7010
    Sample loop, for model 7125, 7010
    Stainless steel
    10 µl
    0,30 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 122,00 €
    122,00 €
    1 * 1 ST
       122,00 €
  • 554-2071
    Sample loop, for model 7125, 7010
    Sample loop, for model 7125, 7010
    Stainless steel
    20 µl
    0,51 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 110,00 €
    110,00 €
    1 * 1 ST
       110,00 €
  • 554-2072
    Sample loop, for model 7125, 7010
    Sample loop, for model 7125, 7010
    Stainless steel
    50 µl
    0,51 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 128,00 €
    128,00 €
    1 * 1 ST
       128,00 €
  • 554-2073
    Sample loop, for model 7125, 7010
    Sample loop, for model 7125, 7010
    Stainless steel
    100 µl
    0,51 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 123,00 €
    123,00 €
    1 * 1 ST
       123,00 €
  • 554-2075
    Sample loop, for model 7125, 7010
    Sample loop, for model 7125, 7010
    Stainless steel
    500 µl
    0,76 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 147,00 €
    147,00 €
    1 * 1 ST
       147,00 €
  • 554-2076
    Sample loop, for model 7125, 7010
    Sample loop, for model 7125, 7010
    Stainless steel
    1000 µl
    0,76 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 169,00 €
    169,00 €
    1 * 1 ST
       169,00 €
  • 554-2078
    Sample loop, for model 7125, 7010
    Sample loop, for model 7125, 7010
    Stainless steel
    5000 µl
    1,00 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 424,00 €
    424,00 €
    1 * 1 ST
       424,00 €