Orders LinkContactUsLinkComponent
Handheld Digital Microscopes, Universal
Handheld Digital Microscopes, Universal
Handheld Digital Microscopes, Universal
Certificates
Handheld Digital Microscopes, Universal
Handheld Digital Microscopes, Universal
16 Options Available Below
Product Details & Documents
The Universal series provides a broad range of products with high image quality to satisfy the most demanding user. Magnification range: 10 to 220×.
  • High speed USB 2.0 connection
  • Metal or composite housing
  • Edge series models with high quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
  • Wireless streaming to a tablet, Smart phone, or computer is possible in combination with WF-10 unit (to be ordered separately)
  • Integrated polariser greatly reduces glare on reflective surfaces (some models only)
Models with Automatic Magnification Reading (AMR): Allows the included software to read the magnification setting from the microscope device, simplifying the measuring process.
Models with Extended Depth of Field (EDOF): Automatically capture and combine multiple images at different focus levels to produce a final image with greater depth of field.
Models with Extended Dynamic Range (EDR): Automatically capture and combine multiple images at different exposure levels to create an image with greater dynamic range.
Models with Flexible LED Control (FLC): Additional LED controls within the included software for adjusting LED brightness and on/off toggle by quadrant.
Delivery information: Supplied with protecting pouch, software CD (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)) and calibration target.
Product Family Options
Models with metal housing (ESD-safe)
Product Information
  • Model
    Magnification
    Illumination
    Resolution
    Additional features
    Availability
    Price
  • 630-1345
    Handheld microscope with polariser
    Handheld microscope with polariser
    UNIVERSAL, AM4013MZT
    10 - 70×, 200×
    8 LEDs
    1,3 MP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 648,00 €
    648,00 €
    1 * 1 ST
       648,00 €
  • 620-2617
    Handheld microscope
    Handheld microscope
    UNIVERSAL, AM4013MT
    10 - 70×, 200×
    8 LEDs
    1,3 MP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 560,00 €
    560,00 €
    1 * 1 ST
       560,00 €
  • 630-2656
    Handheld microscope with exchangeable caps + polariser, Edge
    Handheld microscope with exchangeable caps + polariser, Edge
    UNIVERSAL, AM7115MZT
    20 - 220×
    8 LEDs
    5,0 MP
    FLC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 030,00 €
    1 030,00 €
    1 * 1 ST
       1 030,00 €
  • 630-2658
    Handheld microscope with exchangeable caps + polariser, Edge
    Handheld microscope with exchangeable caps + polariser, Edge
    UNIVERSAL, AM7515MZT
    20 - 220×
    8 LEDs
    5,0 MP
    AMR/FLC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 100,00 €
    1 100,00 €
    1 * 1 ST
       1 100,00 €
  • 630-2752
    Handheld microscope with exchangeable caps + polariser, Edge
    Handheld microscope with exchangeable caps + polariser, Edge
    UNIVERSAL, AM7915MZT
    10 - 220×
    8 LEDs
    5,0 MP
    AMR/EDOF/EDR/FLC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 290,00 €
    1 290,00 €
    1 * 1 ST
       1 290,00 €
  • IDCPAM7115MZTW
    Handheld microscope with exchangeable caps + polariser, Edge
    Handheld microscope with exchangeable caps + polariser, Edge
    UNIVERSAL, AM7115MZTW
    10 - 50×
    8 LEDs
    5,0 MP
    Macro zoom/FLC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 050,00 €
    1 050,00 €
    1 * 1 ST
       1 050,00 €
  • IDCPAM7515MT2A
    Handheld microscope with exchangeable caps, Edge
    Handheld microscope with exchangeable caps, Edge
    UNIVERSAL, AM7515MT2A
    130 - 220×
    9 LEDs
    5,0 MP
    AMR/FLC/Coaxial
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 180,00 €
    1 180,00 €
    1 * 1 ST
       1 180,00 €
Models with composite/plastic housing
Product Information
  • Model
    Magnification
    Illumination
    Resolution
    Additional features
    Availability
    Price
  • 630-1157
    Handheld microscope
    Handheld microscope
    UNIVERSAL, AM4113T
    10 - 70×, 220×
    8 LEDs
    1,3 MP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 384,00 €
    384,00 €
    1 * 1 ST
       384,00 €
  • 630-1165
    Handheld microscope with polariser
    Handheld microscope with polariser
    UNIVERSAL, AM4113ZT
    10 - 70×, 200×
    8 LEDs
    1,3 MP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 472,00 €
    472,00 €
    1 * 1 ST
       472,00 €
  • 620-2618
    Handheld microscope with exchangeable caps, Edge
    Handheld microscope with exchangeable caps, Edge
    UNIVERSAL, AM4115T
    20 - 220×
    8 LEDs
    1,3 MP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 593,00 €
    593,00 €
    1 * 1 ST
       593,00 €
  • 620-2620-
    Handheld microscope with exchangeable caps + polariser, Edge
    Handheld microscope with exchangeable caps + polariser, Edge
    UNIVERSAL, AM4115ZTW
    10 - 50×
    8 LEDs
    1,3 MP
    Macro zoom
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 714,00 €
    714,00 €
    1 * 1 ST
       714,00 €
  • 620-2621
    Handheld microscope with exchangeable caps + polariser, Edge
    Handheld microscope with exchangeable caps + polariser, Edge
    UNIVERSAL, AM4515ZT
    20 - 220×
    8 LEDs
    1,3 MP
    AMR
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 747,00 €
    747,00 €
    1 * 1 ST
       747,00 €
  • 620-2622
    Handheld microscope with exchangeable caps + polariser, Edge
    Handheld microscope with exchangeable caps + polariser, Edge
    UNIVERSAL, AM4815ZT
    20 - 220×
    8 LEDs
    1,3 MP
    EDOF/EDR
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 912,00 €
    912,00 €
    1 * 1 ST
       912,00 €
  • 620-2619
    Handheld microscope with exchangeable caps + polariser, Edge
    Handheld microscope with exchangeable caps + polariser, Edge
    UNIVERSAL, AM4115ZT
    20 - 220×
    8 LEDs
    1,3 MP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 692,00 €
    692,00 €
    1 * 1 ST
       692,00 €
  • IDCPAM4515T
    Handheld microscope with exchangeable caps, Edge, without polariser
    Handheld microscope with exchangeable caps, Edge, without polariser
    UNIVERSAL, AM4515T
    20 - 220×
    8 LEDs
    1,3 MP
    AMR
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 648,00 €
    648,00 €
    1 * 1 ST
       648,00 €
  • IDCPAM4815T
    Handheld microscope with exchangeable caps + without polariser, Edge
    Handheld microscope with exchangeable caps + without polariser, Edge
    UNIVERSAL, AM4815T
    20 - 220×
    8 LEDs
    1,3 MP
    EDOF/EDR
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 642,00 €
    642,00 €
    1 * 1 ST
       642,00 €
    Specifications:
    New Product
    New Product
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Extension pole RK-06-PL, to double the height of the RK-06A / RK-06-AE stand (required when working with Long Working Distance and Extra Long Working Distance microscopes)
    Extension pole RK-06-PL, to double the height of the RK-06A / RK-06-AE stand (required when working with Long Working Distance and Extra Long Working Distance microscopes)
    IDCPRK-06-PL
    Log in for availability
    1 * 1 ST - 28,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Foot pedal, enables users to take a picture with their foot
    Foot pedal, enables users to take a picture with their foot
    630-1821
    Log in for availability
    1 * 1 ST - 63,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Sturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal arm
    Sturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal arm
    630-2517
    Log in for availability
    1 * 1 ST - 340,00 €
Product Information
  • Catalog #
    W×D×H
    Availability
    Price
  • Tabletop stand for handheld Dino-Lite microscopes, RK-06-A, incl. extension arm RK-10-EX
    Tabletop stand for handheld Dino-Lite microscopes, RK-06-A, incl. extension arm RK-10-EX
    IDCPRK-06A
    150×220×270 mm
    Log in for availability
    1 * 1 ST - 252,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Wi-Fi streamer with operating manual, power adapter and 1880 Ah Li-Ion battery
    Wi-Fi streamer with operating manual, power adapter and 1880 Ah Li-Ion battery
    630-2923
    Log in for availability
    1 * 1 ST - 329,00 €
Product Information
  • Catalog #
    W×D×H
    Availability
    Price
  • Tabletop stand for handheld Dino-Lite microscopes, RK-06-AE, with ESD-safe coating, incl. extension arm RK-10-EX
    Tabletop stand for handheld Dino-Lite microscopes, RK-06-AE, with ESD-safe coating, incl. extension arm RK-10-EX
    IDCPRK-06AE
    150×220×270 mm
    Log in for availability
    1 * 1 ST - 249,00 €