Order Entry
Austria
Orders LinkContactUsLinkComponent
Competent cell buffer

Competent cell buffer

Competent cell buffer (60 mM calcium chloride, 15% glycerol, 10 mM tris Hcl, pH 7,5), 100 mL
Catalog # ROCKMB-057
Competent cell buffer
Catalog # ROCKMB-057
CAS Number:  
Competent cell buffer

Competent cell buffer (60 mM calcium chloride, 15% glycerol, 10 mM tris Hcl, pH 7,5), 100 mL

CAS Number:  
Price
239,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

This product is ready-to-use as a working 1X solution and requires no further dilution. This buffer consists of 60 mM Calcium Chloride, 15% v/v Glycerol, 10mM Tris HCl at a pH of 7,5.

Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionised water.

This product was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.

Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.

Documents

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Competent cell buffer (60 mM calcium chloride, 15% glycerol, 10 mM tris Hcl, pH 7,5)

Specifications