Order Entry
Austria
ContactUsLinkComponent
Mag-Bind® cfDNA kit
Mag-Bind® cfDNA kit
Supplier:  OMEGA BIO-TEK
Certificates
undefined
Mag-Bind® cfDNA kit
Supplier:  OMEGA BIO-TEK
Certificates

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    cfDNA
  • Sample type:
    Biofluids (serum, plasma, saliva)
  • Target molecule:
    DNA
  • Sample size:
    0,5 - 4 ml
  • Description:
    Mag-Bind® cfDNA Kit

Specifications

Frequently Bought Together