Order Entry
Austria
ContactUsLinkComponent
Cleanroom mops, MicroKing
Cleanroom mops, MicroKing
  PPSA2140080
undefined
Cleanroom mops, MicroKing
  PPSA2140080

 

  • Description:
    MicroKing, 150 g
  • Length:
    400 mm
  • Pk:
    1

 

 

Mops for grey areas and for all water-resistant hard floor coverings.

  • Increased cleaning performance and maximum dirt absorption facilities
  • Highest area performance and best tuft-quality
  • Made up of synthetic/viscose yarn (50:50) with additional microfiber lines
  • With cover sheet and PVC pocket, sewed up double-latching pocket
  • Ready-to-use, pre-washed

For the use in ISO class 9.