Order Entry
Austria
ContactUsLinkComponent
Cleanroom mops, MicroSicuro trapezoid CR/A
Cleanroom mops, MicroSicuro trapezoid CR/A
Catalog # PPSA2700293
undefined
Cleanroom mops, MicroSicuro trapezoid CR/A
Catalog # PPSA2700293

Specifications

  • Description:
    MicroSicuro rrapezoidal CR/ A, 80 g
  • L×W:
    270×220 mm
  • Pk:
    1

Specifications

About this item

Mops for cleaning surfaces like insulators, and cleaning of hard-to-reach areas. Made up of 100% PES microfibre with soft brush special.

  • Removes 99,9% of all particles in a one-step process
  • Safe disinfection of GMP areas by validating wetting behavior
  • With water-permeable cover sheet polyester bag and drainage function lining
  • Suitable to all mop frames Trapez
  • Autoclavable up to 50 preparation cycles, disinfectant proof and resistant to gamma rays

Ideal for removing disinfectant residues and maximum square footage.