Order Entry
Austria
ContactUsLinkComponent
Cleanroom mops, classic mop
Cleanroom mops, classic mop
Catalog # PPSA2750070
undefined
Cleanroom mops, classic mop
Catalog # PPSA2750070

Specifications

  • Description:
    Classic mop with outer sling, 180 g
  • Length:
    400 mm
  • Pk:
    1

Specifications

About this item

The classic cotton mop with textile pocket.

  • Finely prewashed mops
  • Made up of cotton/poiyester (35:65)