Orders LinkContactUsLinkComponent
Indicator strips, non bleeding
Indicator strips, non bleeding
Indicator strips, non bleeding, 0 - 6,0
Catalog # 315052J
Supplier:   VWR Chemicals
Indicator strips, non bleeding
Catalog # 315052J
Supplier:  VWR Chemicals
Indicator strips, non bleeding
Indicator strips, non bleeding
Indicator strips, non bleeding, 0 - 6,0
Price
13,90 €
Log in for availability
1 * 100 ST
Product Details & Documents
Non bleeding special indicator dyes that are covalently bound onto the reagent paper and then adhered to plastic strips.
Specifications
  • pH range:
    0 - 6,0
  • Description:
    Indicator strips, non bleeding