Reusable half masks, X-plore® 3300 and X-plore® 3500
: Dräger
The X-plore® 3300 and 3500 half masks are low maintenance and affordable. They are equipped with two lateral bayonet connections that enable a whole range of bayonet filters to be used.
- Flexible nose section provides an extra tight seal and excellent fit even when wearing safety goggles
- Side filters and a shallow design, give an exceptional field of vision and X-plore® fits perfectly under protective visors
- The soft FlexiFit head cradle is a perfect fit for all head shapes - without trapping hair; it is also barely noticeable under helmets
- Easily adjustable head strap
X-plore 3500 - special features:
The mask body is made from an innovative, new material: Very kind to the skin, soft and silicone-free; features that combine premium comfort with excellent durability
Drop-down strap option: By unclasping the neck strap, the mask can be removed from your face without taking off your head protection; the inner surface of the mask faces the body, protecting it from contamination
Weight: 95 g
: EN 140
Frequently Bought Together