Cleanroom clipboard TexWrite®
811-0123
: Texwipe
- Description:Cleanroom TexWrite® clipboard, TX5835
- Pk:12
The polypropylene cleanroom clipboard is the ideal portable writing surface for use in critical environments. The rugged clipboard is constructed from rigid polypropylene and features a druable plastic clip mechanism. Rigid back allows use while standing. This clipboard is available for use with any TexWrite® documentation material.
- Chemical resistant, corrosion resistant, durable construction
- Suitable for use with 8,5×11" or A4 paper
- Securely holds 1 to 50 sheets of cleanroom paper
- Rounded corners, no sharp edges to damage gloves or garments
- Cleanroom packaged