Orders LinkContactUsLinkComponent
Conductive grid tapes, CGT™
 :   MICRONOVA
Conductive grid tapes, CGT™
 :  MICRONOVA
Conductive grid tapes, CGT™
Conductive grid tapes, CGT™
Conductive, anti-static polypropylene tape, specially designed with conductive grids embedded between two anti-static layers, for use on devices in ESD sensitive environments.
  • Heavy-duty, anti-static copolymer tape with acrylic adhesive
  • Manufactured for use in ESD sensitive environments
  • Features a conductive grid embedded between two antistatic layers
  • Conducts charge away from substrate
  • Protects sensitive devices that would be otherwise damaged by normal insulating tape
  • Provides an effective static shield and prevents static buildup
  • Prevents shedding, flaking, cracking, or chipping of the grids
  • 36 yards wound on a 3" plastic core
The Micronova CGT™ Conductive Grid Tape features conductive grids that are embedded between two anti-static layers to provide an effective static shield. This design prevents static build-up, as well as preventing any shedding, flaking, cracking, or chipping of the grids.
 : For additional widths, contact your Avantor sales representative.
Product Family Options
  • Description
    Sterile
    Length
    Width
  • Cleanroom conductive grid tape, CGT™
    -
    33 m (36 yds.)
    10 mm
    1 * 1 Rolle
    1 * 1 Rolle - 15,20 €
    15,20 €
    1 * 1 Rolle
       15,20 €
  • Cleanroom conductive grid tape, CGT™
    -
    33 m (36 yds.)
    13 mm
    1 * 1 Rolle
    1 * 1 Rolle - 18,40 €
    18,40 €
    1 * 1 Rolle
       18,40 €
  • Cleanroom conductive grid tape, CGT™
    -
    33 m (36 yds.)
    25 mm
    1 * 1 Rolle
    1 * 1 Rolle - 27,90 €
    27,90 €
    1 * 1 Rolle
       27,90 €
  • Cleanroom conductive grid tape, CGT™
    -
    33 m (36 yds.)
    38 mm
    1 * 1 Rolle
    1 * 1 Rolle - 42,40 €
    42,40 €
    1 * 1 Rolle
       42,40 €
  • Cleanroom conductive grid tape, CGT™
    -
    33 m (36 yds.)
    152 mm
    1 * 1 Rolle
    1 * 1 Rolle - 157,00 €
    157,00 €
    1 * 1 Rolle
       157,00 €
  • Cleanroom conductive grid tape, CGT™
    -
    19,0 mm
  • Cleanroom conductive grid tape, CGT™
    -
    50,8 mm
    1 * 1 Rolle
    1 * 1 Rolle - 53,70 €
    53,70 €
    1 * 1 Rolle
       53,70 €