Anti-HNRNPA2B1 Rabbit Polyclonal Antibody (Cy3®)
BOSSBS-9091R-CY3
: Bioss
- Antibody type:Primary
- Antigen name:Heterogeneous Nuclear Ribonucleoprotein A2/B1
- Antigen symbol:HNRNPA2B1
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:3181
- Antigen synonyms:HNRPA2B1|HNRPA2|HNRPB1|HNRNPA2|SNRPB1|IBMPFD2|RNPA2|HNRNPB1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Involved with pre-mRNA processing. Forms complexes (ribonucleosomes) with at least 20 other different hnRNP and heterogeneous nuclear RNA in the nucleous.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HNRNPA2B1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat