Order Entry
Austria
ContactUsLinkComponent
Anti-GPRIN3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-GPRIN3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-8277R-FITC
Supplier:  Bioss
Anti-GPRIN3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-8277R-FITC
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    G Protein Regulated Inducer Of Neurite Outgrowth 3
  • Antigen symbol:
    GPRIN3
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    285513
  • Antigen synonyms:
    GRIN3|GRIN3_HUMAN.|G protein-regulated inducer of neurite outgrowth 3|Gprin3|GPRIN family member 3
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

May be involved in neurite outgrowth.G protein-coupled receptors (GPCRs) represent a large superfamily of cell-surface receptors that are involved in a multitude of physiological processes such as perception of sensory information, modulation of synaptic transmission, hormone release/action, regulation of cell contraction/migration and cell growth/differentiation. GPCRs interact with G proteins (heterotrimeric GTPases) to synthesize intracellular second messengers, such as diacylglycerol, cyclic AMP, inositol phosphates and calcium ions. Their diverse biological functions range from vision and olfaction to neuronal and endocrine signaling, and are involved in many pathological conditions. GRIN3 (G protein-regulated inducer of neurite outgrowth 3), also known as GPRIN3, is a 776 amino acid protein that contains a C-terminal region which shares a high homology with GRIN2 and GRIN1, and may function in neurite outgrowth.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPRIN3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat