Order Entry
Austria
ContactUsLinkComponent
Anti-IKK alpha + IKK beta Rabbit Polyclonal Antibody (Cy7®)
Anti-IKK alpha + IKK beta Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-7557R-CY7
Supplier:  Bioss
Anti-IKK alpha + IKK beta Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-7557R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    IKK alpha + IKK beta
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

IKK is a serine protein kinase, and the IKK complex contains alpha and beta subunits (IKK alpha and IKK beta). IKK alpha and IKK beta interact with each other and both are essential for NFkB activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IKK alpha + IKK beta
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat