Anti-ANP32B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-6769R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Acidic Nuclear Phosphoprotein 32 Family, Member B
- Antigen symbol:ANP32B
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:10541
- Antigen synonyms:APRIL|PHAPI2|SSP29
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Multifunctional protein working as a cell cycle progression factor as well as a cell survival factor. Required for the progression from the G1 to the S phase. Anti-apoptotic protein which functions as a caspase-3 inhibitor. Has no phosphatase 2A (PP2A) inhibitor activity (By similarity). Exhibits histone chaperone properties, stimulating core histones to assemble into a nucleosome.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ANP32B
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat