Order Entry
Austria
ContactUsLinkComponent
Anti-ANP32B Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-ANP32B Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-6769R-A647
Supplier:  Bioss
Anti-ANP32B Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-6769R-A647
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Acidic Nuclear Phosphoprotein 32 Family, Member B
  • Antigen symbol:
    ANP32B
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    10541
  • Antigen synonyms:
    APRIL|PHAPI2|SSP29
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Multifunctional protein working as a cell cycle progression factor as well as a cell survival factor. Required for the progression from the G1 to the S phase. Anti-apoptotic protein which functions as a caspase-3 inhibitor. Has no phosphatase 2A (PP2A) inhibitor activity (By similarity). Exhibits histone chaperone properties, stimulating core histones to assemble into a nucleosome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANP32B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat