Anti-CDH15 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-6678R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:Cadherin 15
- Antigen symbol:CDH15
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1013
- Antigen synonyms:CDH15|Muscle Cadherin|M-Cadherin|CDH14|Cadherin-14|Cadherin-15|CDH3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:395-442/814
- Purification:Purified by Protein A
- Pk:100 µl
Cadherins are calcium-dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types. M-cadherin is part of the myogenic program and may provide a trigger for terminal muscle differentiation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CDH15
Clonality: Polyclonal
Clone: 0
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: