Anti-DEPDC1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-6525R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:DEP Domain Containing 1
- Antigen symbol:DEPDC1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:55635
- Antigen synonyms:DEPDC1A|cell cycle control protein SDP35|DEP.8|DEPDC1-V2|DEP domain-containing protein 1A|SDP35|5830484J08Rik
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
DEPDC1
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DEPDC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat