Order Entry
Austria
ContactUsLinkComponent
Anti-FSTL1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anti-FSTL1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-6050R-A555
Supplier:  Bioss
Anti-FSTL1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-6050R-A555
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Follistatin-related Protein 1
  • Antigen symbol:
    FSTL1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    11167
  • Antigen synonyms:
    AW107808|FRP|TSC-36|AI316791|FSL1|MIR198
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

TSC-36 is a secreted extracellular glycoprotein. The amino acid sequence of TSC-36 is similar to follistatin, an inhibitor of activin, as it contains a follistatin module. TSC-36 is a heparin-binding protein suggested to have a role in the negative regulation of cellular growth, as its expression is induced in response to TGF-b1. In addition, TSC-36 is not found in small cell lung cancer (SCLC) cells, a highly aggressive neoplasm, but is detected in some non-small cell lung cancer (NSCLC) cells, a moderately aggressive neoplasm.. May modulate the action of some growth factors on cell proliferation and differentiation. Binds heparin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FSTL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat