Anti-BACE Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-5215R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Beta-secretase 1
- Antigen symbol:BACE
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:23621
- Antigen synonyms:Memapsin-2|Beta-site amyloid precursor protein cleaving enzyme 1|BACE1|Membrane-associated aspartic protease 2|ASP2|Asp 2|Beta-secretase 1|Aspartyl protease 2|BACE|Beta-site APP cleaving enzyme 1
- Modification:phospho-Ser498
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Cerebral deposition of amyloid beta peptide is an early and critical feature of Alzheimer's disease. Amyloid beta peptide is generated by proteolytic cleavage of amyloid precursor protein(APP) by two proteases, one of which is the protein encoded by this gene. The encoded protein, a member of the peptidase A1 protein family, is a type I integral membrane glycoprotein and aspartic protease that is found mainly in the Golgi. Multiple transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: BACE
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat