Anti-NT5E Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-4834R-CY3
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:5'-Nucleotidase, Ecto
- Antigen symbol:NT5E
- Clonality:Polyclonal
- Conjugation:Cy3®
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Chicken,Human,Dog,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:4907
- Antigen synonyms:eN|E5NT|NTE|NT5|CALJA|eNT|NT|CD73
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:208-222/574
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Hydrolyzes extracellular nucleotides into membrane permeable nucleosides. Exhibits AMP-, NAD-, and NMN-nucleosidase activities.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200; IF(ICC)(1:50-200
Type: Primary
Antigen: NT5E
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Dog, Chicken