Anti-CDKN2AIP Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-4267R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CDKN2A Interacting Protein
- Antigen symbol:CDKN2AIP
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:55602
- Antigen synonyms:CARF
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
CARF was first cloned as a novel binding partner of ARF from a yeast interactive screen. CARF and ARF colocalize in the perinucleolar region and have a collaborative function. In the nucleoplasm, CARF interacts with p53 and enhances its function. The p53 downregulates CARF in a negative feedback regulatory loop and may also involve p53 antagonist HDM2.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CDKN2AIP
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: