Order Entry
Austria
ContactUsLinkComponent
 
Anti-SERPINB5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-0792R-A350
 :  Bioss
Anti-SERPINB5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-0792R-A350
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Serpin Peptidase Inhibitor, Clade B (ovalbumin), Member 5
  • Antigen symbol:
    SERPINB5
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    5268
  • Antigen synonyms:
    maspin|PI5
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    320-370/375
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Tumor suppressor. It blocks the growth, invasion, and metastatic properties of mammary tumors. As it does not undergo the S (stressed) to R (relaxed) conformational transition characteristic of active serpins, it exhibits no serine protease inhibitory activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SERPINB5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat