Anti-PRDX1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-3875R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Peroxiredoxin 1
- Antigen symbol:PRDX1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5052
- Antigen synonyms:MSP23|PAGA|PAG|thioredoxin-dependent peroxide reductase 2|PRDX1|peroxiredoxin 1|PRXI|NKEFA|natural killer-enhancing factor A|proliferation-associated gene A|PRX1|TDPX2|PAGB
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:63-88/199
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Involved in redox regulation of the cell. Reduces peroxides with reducing equivalents provided through the thioredoxin system but not from glutaredoxin. May play an important role in eliminating peroxides generated during metabolism. Might participate in the signaling cascades of growth factors and tumor necrosis factor-alpha by regulating the intracellular concentrations of H(2)O(2). Reduces an intramolecular disulfide bond in GDPD5 that gates the ability to GDPD5 to drive postmitotic motor neuron differentiation (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PRDX1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat