Anti-EPHA10 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-10052R-A488
: Bioss
- Antibody type:Primary
- Antigen name:EPH Receptor A10
- Antigen symbol:EPHA10
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:284656
- Antigen synonyms:ephrin type-A receptor 10|EphA10s protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:150-200/1008
- Purification:Purified by Protein A
- Pk:100 µl
Receptor for members of the ephrin-A family. Binds to EFNA3, EFNA4 and EFNA5.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: EPHA10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat