Order Entry
Austria
ContactUsLinkComponent
 
Anti-NOX5 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-10156R-A647
 :  Bioss
Anti-NOX5 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-10156R-A647
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    NAPDH oxidase 5
  • Antigen symbol:
    NOX5
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    339345
  • Antigen synonyms:
    NADPH oxidase EFhand Calcium binding domain 5|NOX5|NOX5_HUMAN|MGC149776|MGC149777.|NADPH oxidase 5
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Calcium-dependent NADPH oxidase that generates superoxide. Also functions as a calcium-dependent proton channel and may regulate redox-dependent processes in lymphocytes and spermatozoa. May play a role in cell growth and apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NOX5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human