Specifications
- Antibody type:Primary
- Antigen name:Glutamate Receptor 1
- Antigen symbol:GRIA1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2890
- Antigen synonyms:GLUH1|AMPA 1|AMPA-selective glutamate receptor 1|GLURA|GLUR1|glutamate receptor 1|gluR-A|GluA1|MGC133252|gluR-1|HBGR1|gluR-K1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Glutathione reductase (GR) is a member of pyridine nucleotide- disulfideoxidoreductases, which includes the closely related enzymes thioredoxin reductase, lipoamide dehydrogenase, trypanothione reductase and mercuric ion reductase. GR is a cytoplasmic flavoenzyme widely distributed in aerobic organisms. The dimeric protein is composed of two identical subunits, each containing 1 FAD and 1 redox-active disulfide/dithiol as components of the catalytic apparatus. It plays a role in maintaining glutathione (GSH) in its reduced form by catalyzing the reduction of glutathione disulfide (GSSG): GSSG + NADPH + H+?2GSH + NADP+. In mosteukaryotic cells, GR maintains the ratio of [GSH]/[GSSG], and participates in several vital functions such as the detoxification of reactive oxygen species as well as protein and DNA biosynthesis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GLUR/glutathione reductase
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat