Anti-IL3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-2598R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:Interleukin 3
- Antigen symbol:IL3
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:16187
- Antigen synonyms:Multipotential colony-stimulating factor|P-cell-stimulating factor|MCGF|Hematopoietic growth factor|Mast-cell growth factor|Interleukin-3|IL3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:100-150/166
- Purification:Purified by Protein A
- Pk:100 µl
Granulocyte/macrophage colony-stimulating factors are cytokines that act in hematopoiesis by controlling the production, differentiation, and function of 2 related white cell populations of the blood, the granulocytes and the monocytes-macrophages. This CSF induces granulocytes, macrophages, mast cells, stem cells, erythroid cells, eosinophils and megakaryocytes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IL3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat