Anti-TNFA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-2081R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Alpha
- Antigen symbol:TNFA
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:21926
- Antigen synonyms:tumor necrosis factor ligand superfamily member 2|tumor necrosis factor alpha|DIF|monocyte-derived|Tnfa|cachectin|Tnfsf1a|TNFalpha|member 2|TNF-alpha|TNF-a|TNFSF2|macrophage-derived|TNF superfamily|TNF|tumor necrosis factor-alpha|APC1 protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:180-230/235
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Cytokine that binds to TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. It is mainly secreted by macrophages and can induce cell death of certain tumor cell lines. It is potent pyrogen causing fever by direct action or by stimulation of interleukin-1 secretion and is implicated in the induction of cachexia, Under certain conditions it can stimulate cell proliferation and induce cell differentiation. The TNF intracellular domain (ICD) form induces IL12 production in dendritic cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TNF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat