Anti-PRDX3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-1874R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Peroxiredoxin 3
- Antigen symbol:PRDX3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:10935
- Antigen synonyms:AOP1|MGC24293|AOP-1|MGC104387|SP-22|PRO1748|MER5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:90-130/256
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Involved in redox regulation of the cell. Protects radical-sensitive enzymes from oxidative damage by a radical-generating system. Acts synergistically with MAP3K13 to regulate the activation of NF-kappa-B in the cytosol.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PRDX3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat