Anti-INHBA Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-1774R-CY3
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Inhibin, Beta A
- Antigen symbol:INHBA
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:3624
- Antigen synonyms:FRP|EDF
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:340-385/426
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Inhibins and activins inhibit and activate, respectively, the secretion of follitropin by the pituitary gland. Inhibins/activins are involved in regulating a number of diverse functions such as hypothalamic and pituitary hormone secretion, gonadal hormone secretion, germ cell development and maturation, erythroid differentiation, insulin secretion, nerve cell survival, embryonic axial development or bone growth, depending on their subunit composition. Inhibins appear to oppose the functions of activins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: INHBA
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat