Specifications
- Antibody type:Primary
- Antigen name:Leucine Rich Repeat Containing 6
- Antigen symbol:LRRC6
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:LRTP|TSLRP|CILD19
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The leucine-rich repeat-containing protein 6 (LRRC6), also designated Leucine-rich testis-specific protein (LRTP), is a 466 amino acid protein that contains 3 LRR repeats and plays a role in spermatogenesis. The gene encoding LRRC6 maps to chromosome 8, which encodes approximately 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukemias and lymphomas, and typically associated with a poor prognosis. Chromosome 8 is also associated with Trisomy 8, Pfeiffer syndrome, congenital hypothyroidism and Waardenburg syndrome.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LRRC6
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat