Specifications
- Antibody type:Primary
- Antigen name:Polypeptide N-Acetylgalactosaminyltransferase 13
- Antigen symbol:GALNT13
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:114805
- Antigen synonyms:GalNAc-T13
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes are substrate-specific proteins that catalyze the transfer of GalNAc (N-acetylgalactosamine) to serine and threonine residues onto various proteins, thereby initiating mucin-type O-linked glycosylation in the Golgi apparatus. GalNAc-T13 (Polypeptide N-acetylgalactosaminyltransferase 13), also known as UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 13, is a 556 amino acid protein that displays much stronger enzymatic activity than GalNAc-1 towards GalNAc transfer to mucin peptides such as Muc5a and Muc7. The N-terminal domain is involved in substrate binding and manganese coordination, while the C-terminal domain is involved in UDP-Gal binding and catalytic reaction. With specific expression in the central nervous system, GalNAc-T13 may be responsible for the synthesis of Tn antigen in neuronal cells, which is a universal carcinoma marker on malignant cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GALNT13/GalNAc-T13
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat