Specifications
- Antibody type:Primary
- Antigen name:Fucose-1-Phosphate Guanylyltransferase
- Antigen symbol:FPGT
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:8790
- Antigen synonyms:GFPP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Guanylyltransferase enzymes transfer one molecule of GTP to another molecule and also function in the transfer of guanosine nucleotides to sugar molecules. The carbohydrate moieties that are generated are covalently attached to cell surfaces and are necessary to ensure a surface contour that satisfies a variety of physiological roles. L-fucose is an important sugar in complex carbohydrates that is frequently found on plant and mammalian N-linked glycans. FPGT (Fucose-1-phosphate guanylyltransferase), also known as GFPP (GDP-L-fucose pyrophosphorylase), is a 594 amino acid cytoplasmic protein that catalyzes the formation of GDP-L-fucose from L-fucose-1-phosphate and GTP. FPGT functions to reutilize the L-fucose that is produced uopn glycoprotein and glycolipid turnover.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FPGT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat